09216 Fluka

Viscosity and Density Standard

N4000, UKAS ISO/IEC17025 and ISO Guide 34 certified, viscosity 10296 mPa.s (25 °C)

DOWNLOAD MSDS (PDF)

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Application Areas and Market Segments, Petrochemistry, Pharma,
shelf life   (limited shelf life, expiry date on the label)
mfr. no.   Paragon Scientific Ltd.
viscosity   10296 mPa.s(25 °C)
density   0.8855 g/mL at 25 °C

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?