09332 Fluka

Aminopyrazine

purum, ≥97.0% (NT)

DOWNLOAD MSDS (PDF)

Synonym: Pyrazinamine

  • CAS Number 5049-61-6

  • Empirical Formula (Hill Notation) C4H5N3

  • Molecular Weight 95.10

  •  Beilstein Registry Number 107025

  •  EC Number 225-748-7

  •  MDL number MFCD00006137

  •  PubChem Substance ID 24846315

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Pyrazines
grade   purum
assay   ≥97.0% (NT)
mp   118-120 °C(lit.)
  118-120 °C

Price and Availability

Customers Also Viewed

Aminopyrazine

matrix substance for MALDI-MS, ≥99.0% (NT)

Aminopyrazine

98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?