09870 Fluka

Ammonium hydroxide solution

purum, ~24% NH3 in H2O (T)

DOWNLOAD MSDS (PDF)

Synonym: Ammonia aqueous, Ammonia water

  • CAS Number 1336-21-6

  • Linear Formula NH4OH

  • Molecular Weight 35.05

  •  Beilstein Registry Number 3587154

  •  EC Number 215-647-6

  •  MDL number MFCD00066650

  •  PubChem Substance ID 24846477

Properties

Related Categories Chemical Synthesis, Inorganic Bases, Synthetic Reagents More...
grade   purum
expl. lim.   27 %
concentration   ~24% NH3 in H2O (T)
density   0.91 g/mL at 20 °C
  0.9 g/mL at 25 °C(lit.)

Description

Other Notes

Sales restrictions may apply

Price and Availability

Customers Also Viewed

Ammonium hydroxide solution

28% NH3 in H2O, ≥99.99% trace metals basis

Ammonium hydroxide solution

ACS reagent, 28.0-30.0% NH3 basis

Ammonium hydroxide solution

ACS reagent, 28.0-30.0% NH3 basis

Ammonium hydroxide solution

puriss. p.a. plus, ≥25% NH3 in H2O

Ammonium hydroxide solution

≥25% in H2O, eluent additive for LC-MS

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?