11093 Fluka

Arsenobetaine

purum p.a., ≥95.0% (NMR)

DOWNLOAD MSDS (PDF)

Synonym: 2-(Trimethylarsonio)acetate

  • CAS Number 64436-13-1

  • Empirical Formula (Hill Notation) C5H11AsO2

  • Molecular Weight 178.06

  •  Beilstein Registry Number 3933180

  •  MDL number MFCD01311729

  •  PubChem Substance ID 24846866

Properties

Related Categories Arsenic Compounds, Building Blocks, Chemical Synthesis, Organic Building Blocks
grade   purum p.a.
assay   ≥95.0% (NMR)

Description

Packaging

50 mg in glass bottle

Price and Availability

Customers Also Viewed

Arsenic(III) oxide

99.995% trace metals basis

Arsenobetaine Standard solution

BCR® certified Reference Material

Cacodylic acid

≥99.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?