Synonym: BTMABr3, Benzyltrimethylammonium bromide dibromide
CAS Number 111865-47-5
Linear Formula C6H5CH2N(Br3)(CH3)3
Molecular Weight 389.95
Beilstein Registry Number 7105839
MDL number MFCD00134423
Popular Documents: FTNMR (PDF)
| Related Categories | Ammonium, Ammonium Salts, Bromination, C-X Bond Formation (Halogen), Chemical Synthesis, |
| grade | purum |
| assay | ≥97.0% (AT) |
| mp | 99-101 °C(lit.) |
| 99-101 °C |
Convenient-to-handle, solid brominating reagent with a greater brominating potential than tetrabutylammonium tribromide. BTMABr3 has been used for the bromination of phenols, methyl ketones, and aromatic amines 1; Hofmann degradation of amides2; Reagent for oxidations3
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?