Synonym: BSA
CAS Number 10416-59-8
Linear Formula CH3C[=NSi(CH3)3]OSi(CH3)3
Molecular Weight 203.43
Beilstein Registry Number 1306669
EC Number 233-892-7
MDL number MFCD00008270
Popular Documents: FTIR (PDF)
| Related Categories | Analytical Reagents, Analytical/Chromatography, Chemical Synthesis, Derivatization Reagents, Derivatization Reagents GC, |
| grade | purum |
| assay | ≥95.0% (GC) |
| refractive index |
n |
|
n | |
| bp | 71-73 °C/35 mmHg(lit.) |
| density | 0.832 g/mL at 20 °C(lit.) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?