Synonym: 17α-20,29,30-Trinorlup-13(18)-ene
CAS Number 63543-60-2
Empirical Formula (Hill Notation) C27H44
Molecular Weight 368.64
EC Number 208-759-1
MDL number MFCD02683736
PubChem Substance ID 24850050
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chemical Class, Hopanes & Gammaracenes, |
| grade | analytical standard |
| assay | ≥97.0% (GC/MS) |
| shelf life | (limited shelf life, expiry date on the label) |
| concentration | 0.05 mg/mL in isooctane |
| storage temp. | 2-8°C |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?