Synonym: Hydrochloric acid in methanol
CAS Number 132228-87-6
Linear Formula HCl · CH3OH
Molecular Weight 36.46
Beilstein Registry Number 1098214
MDL number MFCD00011324
PubChem Substance ID 24850807
Popular Documents: Specification Sheet (PDF)
| Related Categories | Analytical Reagents, Analytical/Chromatography, Derivatization Reagents, Derivatization Reagents GC, Reagents for Alkylation & Esterification More... |
| grade | for GC derivatization |
| concentration | ~1.25 M HCl |
| density | 0.831 g/mL at 20 °C |
| storage temp. | 2-8°C |
Derivatizing agent for GC
Customers Also Viewed
0.5 N
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?