18087 Fluka

(±)-3,4-Methylenedioxymethamphetamine hydrochloride solution

~1 mg/mL in methanol, analytical standard, for drug analysis

DOWNLOAD MSDS (PDF)

Synonym: DL-3,4-Methylenedioxymethamphetamine hydrochloride solution, DL-MDMA hydrochloride solution, XTC hydrochloride solution

  • CAS Number 64057-70-1

  • Empirical Formula (Hill Notation) C11H15NO2 · HCl

  • Molecular Weight 229.70

  •  MDL number MFCD00079444

Properties

Related Categories Alphabetical Index of Analytical Standards, Amphetamine, Analytical Standards, Analytical/Chromatography, Chemical Structure,
grade   analytical standard, for drug analysis
assay   ≥98.5% (HPLC)
shelf life   (limited shelf life, expiry date on the label)
drug control   USDEA Schedule I; regulated under CDSA - not available from Sigma-Aldrich Canada
concentration   ~1 mg/mL in methanol
suitability   corresponds for H-NMR
storage temp.   2-8°C

Description

Packaging

ampoule contains 1.2mL solution

Price and Availability

Customers Also Viewed

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?