18795 Fluka

Brucella Agar

for microbiology

DOWNLOAD MSDS (PDF)

Properties

Related Categories A - M, All Dairy Media (alphabetical), All Food Control Media (alphabetical), All Solid Media/Agars, Brucella,
grade   for microbiology
shelf life   (limited shelf life, expiry date on the label)
composition   agar, 13 g/L
  casein peptone, 10 g/L
  D(+)-glucose, 1 g/L
  meat peptone, 10 g/L
  sodium chloride, 5 g/L
  yeast extract, 2 g/L
final pH   7.0±0.2 (25 °C)
suitability   Haemophilus species
  Pneumococcus
  nonselective for Brucella
  nonselective for Listeria
  nonselective for Neisseria
  nonselective for Streptococcus

Description

Application

For the isolation and cultivation of Brucella species, in particular the pathogens B. melitensis, B. abortus and B. suis, from clinical material and comestibles of animal origin, modified according to Wundt.

Price and Availability

Customers Also Viewed

for microbiology

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?