19718 Fluka

Plate Count Agar, Vegitone

for microbiology

DOWNLOAD MSDS (PDF)

Synonym: Vegitone Plate Count Agar

Properties

Related Categories All Dairy Media (alphabetical), All Food Control Media (alphabetical), All Solid Media/Agars, Bacillus, Bacteria (General Media),
grade   for microbiology
shelf life   (limited shelf life, expiry date on the label)
composition   agar, 15.0 g/L
  dextrose, 1.0 g/L
  tryptone (vegetable), 5.0 g/L
  yeast extrakt, 2.5 g/L
final pH   7.0±0.2 (25 °C)
suitability   nonselective for Bacillus
  nonselective for Bacteria (General Media)
  nonselective for Escherichia coli and Coliforms
  nonselective for Lactobacillus
  nonselective for Lactococci
  nonselective for Listeria
  nonselective for Staphylococcus
  nonselective for Streptococcus

Description

Application

Vegitone Plate Count Agar medium is recommended for the plate count of microorganisms in foods, water and waste water. In this medium the animal derived peptone is replaced by a plant peptone.

Price and Availability

Customers Also Viewed

for microbiology

for microbiology

for microbiology, ready to use media in the bottle

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?