Synonym:
(R)
CAS Number 6485-40-1
Empirical Formula (Hill Notation) C10H14O
Molecular Weight 150.22
Beilstein Registry Number 2206714
EC Number 229-352-5
MDL number MFCD00001578
PubChem Substance ID 24853181
Popular Documents: FTNMR (PDF)
| Related Categories | A - D, A-F, All Alphabetically Sorted, Alphabetical, Alphabetical Index of Analytical Standards, |
| grade | analytical standard, for terpene analysis |
| assay | ≥99.0% (sum of enantiomers, GC) |
| optical activity | [α]20/D −61.5±2°, neat |
| shelf life | (limited shelf life, expiry date on the label) |
| refractive index |
n |
| bp | 228-230 °C(lit.) |
| density | 0.960 g/mL at 20 °C(lit.) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?