239089 Fluka

Buffer, Hydrion®

pH 7.00±0.02

DOWNLOAD MSDS (PDF)

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Buffer Tablets & Powders, Titration, pH Buffers for Titration More...
vapor density   4.9 (vs air)
packaging   pkg of 5 × (envelopes)
pH   7.00±0.02

Description

General description

Visit our Titration Center to learn more.

Packaging

Envelope is triple-layered to keep buffer completely dry and free-flowing.

Reconstitution

Precise buffer solution is prepared by dissolving contents of envelope in 500 mL distilled water.

Legal Information

Hydrion is a registered trademark of Micro Essential Laboratory, Inc.

Price and Availability

Customers Also Viewed

for 100 mL solution

pH 2.00±0.02

pH 4.00±0.02

pH 10.00±0.02

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?