CAS Number 7790-94-5
Linear Formula ClSO3H
Molecular Weight 116.52
EC Number 232-234-6
MDL number MFCD00011523
PubChem Substance ID 24855890
| Related Categories | Analytical Reagents, Analytical/Chromatography, Chemical Synthesis, Derivatization Reagents, Derivatization Reagents TLC, |
| Quality Level | PREMIUM |
| vapor density | 4 (vs air) |
| vapor pressure | 1 mmHg ( 25 °C) |
| 3.3 mmHg ( 37.7 °C) | |
| grade | purum |
| assay | ≥98.0% (T) |
| expl. lim. | 37.7 % |
may contain particles and discolor to slightly greenish-yellow on storage
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?