32791 Fluka

1,4-Diaminobutane

purum, ≥98.0% (GC)

DOWNLOAD MSDS (PDF)

Synonym: 1,4-Butanediamine, Putrescine, Tetramethylenediamine

  • CAS Number 110-60-1

  • Linear Formula NH2(CH2)4NH2

  • Molecular Weight 88.15

  •  Beilstein Registry Number 605282

  •  EC Number 203-782-3

  •  MDL number MFCD00008235

  •  PubChem Substance ID 24859703

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Bioactive Small Molecules, Building Blocks, Cell Biology, Chemical Synthesis, D-DIF,
grade   purum
assay   ≥98.0% (GC)
expl. lim.   9.08 %
impurities   ≤0.5% water
refractive index   n20/D 1.457(lit.)
bp   158-160 °C(lit.)
mp   25-28 °C(lit.)
density   0.877 g/mL at 25 °C(lit.)
Gene Information   rat ... Odc1(24609)

Price and Availability

Customers Also Viewed

1,4-Diaminobutane

puriss., ≥99.0% (GC)

1,4-Diaminobutane dihydrochloride

purum, ≥99.0% (AT)

Cadaverine

purum, ≥97.0% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?