Synonym: NP-1EO technical
EC Number 200-662-2
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, N, NJ - NZ More... |
| grade | analytical standard, for environmental analysis |
| technical | |
| concentration | 5 μg/mL in acetone |
| storage temp. | −20°C |
Certan Vial
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?