Synonym: 3α-Acetoxy-7α,15-dihydroxy-12,13-epoxytrichothec-9-en-8-one, 3α-Acetylvomitoxin, 3-AcDON
CAS Number 50722-38-8
Empirical Formula (Hill Notation) C17H22O7
Molecular Weight 338.35
MDL number MFCD00135964
| Related Categories | All alphabetically sorted, Analytical Standards, Analytical/Chromatography, Biotoxins, Chromatography, |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?