33665 Fluka

Buffer solution pH 4.0 (20 °C)

red colored, citric acid / sodium hydroxide / sodium chloride solution, with fungicide

DOWNLOAD MSDS (PDF)

Properties

Related Categories Analytical Reagents, Analytical Standards, Analytical/Chromatography, Biological Buffers, Buffer Convenience Packaging,
quality   red colored
  with fungicide
pH   4.00 (20 °C)
  4.00 (30 °C)
  4.00 (40 °C)
  4.00 (50 °C)
  4.00 (60 °C)
  4.00 (70 °C)
  4.00 (80 °C)
  4.00 (90 °C)
  4.02 (10 °C)
  4.03 (0 °C)

Description

General description

This standard solution is tested against certified reference materials from NIST and/or PTB.

Visit our Titration Center to learn more.

Packaging

The suffix -VP or -VOLPAC in the product number indicates packaging in 5 or 10 L VOLPAC containers.

Price and Availability

Customers Also Viewed

violet colored, borax / sodium hydroxide

with fungicide, citric acid / sodium hydroxide / sodium chloride solution, traceable to SRM from NIST

potassium dihydrogen phosphate / disodium hydrogen phosphate, with fungicide, traceable to SRM from NIST

green colored, potassium dihydrogen phosphate / disodium hydrogen phosphate, with fungicide

Buffer solution pH 9.0 (20 °C)

blue colored, borax / hydrochloric acid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?