Synonym:
S-Benzyl 1,2-
CAS Number 85785-20-2
Empirical Formula (Hill Notation) C15H23NOS
Molecular Weight 265.41
MDL number MFCD00210288
PubChem Substance ID 24860597
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chromatography, E, |
| grade | PESTANAL®, analytical standard |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?