Synonym:
N-
CAS Number 140-40-9
Empirical Formula (Hill Notation) C5H5N3O3S
Molecular Weight 187.18
EC Number 205-414-7
MDL number MFCD00022438
PubChem Substance ID 24860678
| Related Categories | A, AA to AL, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography More... |
| grade | VETRANAL™, analytical standard |
VETRANAL is a trademark of Sigma-Aldrich Co. LLC
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?