Synonym:
4-
CAS Number 2635-10-1
Empirical Formula (Hill Notation) C11H15NO3S
Molecular Weight 241.31
MDL number MFCD00155393
PubChem Substance ID 24860992
| Related Categories | Acaricides, All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, |
| grade | PESTANAL®, analytical standard |
| storage temp. | 2-8°C |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?