CAS Number 7758-01-2
Linear Formula KBrO3
Molecular Weight 167.00
MDL number MFCD00011359
PubChem Substance ID 24860719
| Related Categories | Analytical Reagents, Analytical/Chromatography, Chemical Synthesis, Halogenated Solutions, Inorganic Salts, |
| quality | volumetric |
| shelf life | (limited shelf life, expiry date on the label) |
| concentration |
|
| factor | 1.000 ±0.002 |
Visit our Titration Center to learn more.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?