Synonym: Mercury(II) nitrate
CAS Number 7783-34-8
Linear Formula Hg(NO3)2
Molecular Weight 324.60
MDL number MFCD00011038
PubChem Substance ID 24861085
| Related Categories | Analytical Reagents, Analytical/Chromatography, G - M, Salt Solutions, Titration, |
| quality | for the determination of chloride |
| volumetric | |
| shelf life | (limited shelf life, expiry date on the label) |
| concentration | 0.05 M Hg(NO3)2 (0.1N) |
| factor | 1.000 ±0.005 |
Visit our Titration Center to learn more.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?