CAS Number 3204-27-1
Empirical Formula (Hill Notation) C12H14N2O6
Molecular Weight 282.25
Beilstein Registry Number 2163325
EC Number 221-706-7
MDL number MFCD00129667
PubChem Substance ID 24861153
| Related Categories | All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chromatography, |
| grade | PESTANAL®, analytical standard |
Restricted to professional users. Attention − Avoid exposure − obtain special instructions before use.
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?