MDL number MFCD00011047
PubChem Substance ID 24861383
| Related Categories | Analytical Reagents, Analytical/Chromatography, G - M, Most common compounds for volumetric titration, Potable- & Waste-Water Testing, |
| grade | for COD determination according to DIN 38409-H43-1 |
| assay | 195-205 g/L |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?