34675 Fluka

Water with 0.1% acetic acid

LC-MS CHROMASOLV®

DOWNLOAD MSDS (PDF)
  • Empirical Formula (Hill Notation) H2O

  • Molecular Weight 18.02

  •  EC Number 231-791-2

  •  MDL number MFCD00011332

  •  PubChem Substance ID 24861436

Properties

Related Categories Amber Glass Bottles, Analytical Reagents, Analytical/Chromatography, Blends- CHROMASOLV LC-MS, Chromatography Reagents & Solvents,
grade   LC-MS CHROMASOLV®
shelf life   (limited shelf life, expiry date on the label)
contains   0.093-0.107 % (w/w) acetic acid as additive
transmittance   210 nm, ≥20%
  230 nm, ≥75%
  254 nm, ≥99%
pH   3.2-3.4 (20 °C)
cation traces   Ca: ≤0.5 mg/kg
  K: ≤0.5 mg/kg
  Mg: ≤0.5 mg/kg
  Na: ≤2 mg/kg
abs.   HPLC-gradient/210 nm ≤50 mAU
  HPLC-gradient/254 nm ≤10 mAU
suitability   LC-MS tested

Description

Packaging

2.5 L in glass bottle

Legal Information

CHROMASOLV is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH

Price and Availability

Customers Also Viewed

Acetic acid

eluent additive for LC-MS

LC-MS CHROMASOLV®

Water
39253Water

LC-MS CHROMASOLV®

LC-MS CHROMASOLV®

Water with 0.1% formic acid

LC-MS CHROMASOLV®

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?