EC Number 200-835-2
MDL number MFCD00001878
PubChem Substance ID 24861437
| Related Categories | Analytical Reagents, Analytical/Chromatography, Chromatography Reagents & Solvents, LC-MS Blends, LC-MS Solvents & Reagents (CHROMASOLV) More... |
| grade | LC-MS CHROMASOLV® |
| assay | ≥99.5% (GC) |
| impurities | 0.009-0.011% trifluoroacetic acid |
| 0.009-0.107% formic acid | |
| transmittance | 210 nm, ≥5% |
| 230 nm, ≥15% | |
| 254 nm, ≥90% |
2.5 L in glass bottle
CHROMASOLV is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?