34807 Fluka

HYDRANAL®-Coulomat A

reagent for coulometric KF titration (anolyte solution), for cells with diaphragm

DOWNLOAD MSDS (PDF)

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Coulometric Reagents, KF Reagents for Special Applications, Karl Fischer Titration Reagents (HYDRANAL),
composition   methanol
  sulfur dioxide
  trichloromethane
suitability   use tested
compatibility   reagent for coulometric KF titration (anolyte solution)

Description

Application

To be used with: see related products

General description

Visit our HYDRANAL center to learn more.

Legal Information

HYDRANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH

Price and Availability

Customers Also Viewed

reagent for volumetric one-component KF titration (methanol-free titrating agent)

reagent for coulometric KF titration (anolyte solution), for cells without diaphragm

reagent for coulometric KF titration (anolyte solution), for cells with and without diaphragm

reagent for coulometric KF titration in long-chained hydrocarbons (anolyte solution), for cells with and without diaphragm

reagent for coulometric KF titration (catholyte solution)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?