34836 Fluka

HYDRANAL®-Coulomat AG

reagent for coulometric KF titration (anolyte solution), for cells with and without diaphragm

DOWNLOAD MSDS (PDF)

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Coulometric Reagents, Karl Fischer Titration Reagents (HYDRANAL), Titration More...
composition   diethanolamine
  imidazole
  methanol
  sulfur dioxide
suitability   use tested
compatibility   reagent for coulometric KF titration (anolyte solution)

Description

Frequently Asked Questions

Live Chat and Frequently Asked Questions are available for this Product.

Application

To be used with: see related products

General description

Visit our HYDRANAL center to learn more.

Legal Information

HYDRANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH

Price and Availability

Customers Also Viewed

reagent for volumetric one-component KF titration (methanol-free titrating agent)

reagent for coulometric KF titration (anolyte solution), for cells with diaphragm

reagent for coulometric KF titration in long-chained hydrocarbons (anolyte solution), for cells with and without diaphragm

reagent for coulometric KF titration with oven (anolyte solution), for cells with and without diaphragm

reagent for coulometric KF titration (catholyte solution)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?