EC Number 200-659-6
MDL number MFCD00004595
PubChem Substance ID 24861705
| Related Categories | Analytical Reagents, Analytical/Chromatography, Chromatography Reagents & Solvents, LC-MS Blends, LC-MS Solvents & Reagents (CHROMASOLV) More... |
| grade | LC-MS CHROMASOLV® |
| assay | ≥99.5% (GC) |
| transmittance | 210 nm, ≥5% |
| 230 nm, ≥35% | |
| 254 nm, ≥90% | |
| cation traces | Ca: ≤0.5 mg/kg |
| K: ≤0.5 mg/kg |
2.5 L in glass bottle
CHROMASOLV is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?