CAS Number 1310-73-2
Linear Formula NaOH
Molecular Weight 40.00
MDL number MFCD00003548
PubChem Substance ID 24861783
| Related Categories | Analytical Reagents, Analytical/Chromatography, Base Solutions, By Reference Material, Essential Chemicals, |
| vapor pressure | 3 mmHg ( 37 °C) |
| quality | volumetric |
| concentration | 8 M NaOH (8N) |
| factor | 1.000 ±0.2% |
Visit our Titration Center to learn more.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?