CAS Number 133855-98-8
Empirical Formula (Hill Notation) C17H13ClFN3O
Molecular Weight 329.76
Beilstein Registry Number 9509539
EC Number 406-850-2
PubChem Substance ID 24862909
| Related Categories | All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chromatography, |
| grade | PESTANAL®, analytical standard |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?