Synonym: Ti(acac)2OiPr2
CAS Number 17927-72-9
Linear Formula [(CH3)2CHO]2Ti(C5H7O2)2
Molecular Weight 364.26
MDL number MFCD00000031
PubChem Substance ID 24863663
Popular Documents: FTIR (PDF)
| Related Categories | Catalysis and Inorganic Chemistry, Chemical Synthesis, Titanium More... |
| grade | purum |
| concentration | ~75% in isopropanol |
| density | 1.00 g/mL at 20 °C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?