CAS Number 1017-88-5
Linear Formula (C6H5)2P(I)(CH3)2
Molecular Weight 342.16
Beilstein Registry Number 3637544
MDL number MFCD00031576
Popular Documents: FTNMR (PDF)
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chemical Synthesis, D, |
| grade | purum |
| assay | ≥98.0% (AT) |
| ign. residue | ≤2.0% |
| mp | 246-249 °C(lit.) |
| 246-251 °C |
The corresponding di-ylide is a powerful reagent for the methylenation of carbonyl compounds1; Ylide and di-ylide are used as ligands for organometallic compounds2,3
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?