Empirical Formula (Hill Notation) I2
Molecular Weight 253.81
Beilstein Registry Number 3587194
MDL number MFCD00011355
PubChem Substance ID 24865312
Popular Documents: Specification Sheet (PDF)
| Related Categories | Analytical Reagents, Analytical/Chromatography, G - M, Halogenated Solutions, Iodine and Iodide solutions, |
| quality | volumetric |
| concentration | 0.025 M I2 (0.05N) |
| density | 1.03 g/mL at 25 °C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?