| Related Categories | Analytical Standards, Analytical/Chromatography, Application Areas and Market Segments, Food and Beverage, Petrochemistry, |
| shelf life | (limited shelf life, expiry date on the label) |
| mfr. no. | Paragon Scientific Ltd. |
| viscosity | 72439 mPa.s(25 °C) |
| density | 0.8951 g/mL at 25 °C |
Customers Also Viewed
N350, UKAS ISO/IEC17025 and ISO Guide 34 certified, viscosity 740.3 mPa.s (25 °C)
N15000, UKAS ISO/IEC17025 and ISO Guide 34 certified, viscosity 40006 mPa.s (25 °C)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?