Synonym:
2-Chloro-2′,6′-diethyl-N-
CAS Number 15972-60-8
Empirical Formula (Hill Notation) C14H20ClNO2
Molecular Weight 269.77
Beilstein Registry Number 2944476
EC Number 240-110-8
MDL number MFCD00041817
PubChem Substance ID 24868764
| Related Categories | 2000/60/EC, A, A-B, AA to AL, All alphabetically sorted, |
| grade | PESTANAL®, analytical standard |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?