Synonym: γ-BHC, 1α,2α,3β,4α,5α,6β-Hexachlorocyclohexane
CAS Number 58-89-9
Empirical Formula (Hill Notation) C6H6Cl6
Molecular Weight 290.83
Beilstein Registry Number 1907337
EC Number 200-401-2
MDL number MFCD00135947
PubChem Substance ID 24869073
| Related Categories | Acaricides, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Application Areas and Market Segments, |
| grade | PESTANAL®, analytical standard |
| mp | 113-115 °C(lit.) |
| Gene Information |
mouse ... Esr1(13982) rat ... Ar(24208) |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?