Synonym:
2,4-
CAS Number 1610-18-0
Empirical Formula (Hill Notation) C10H19N5O
Molecular Weight 225.29
Beilstein Registry Number 613574
EC Number 216-548-0
MDL number MFCD00047343
PubChem Substance ID 24869164
| Related Categories | All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chromatography, |
| grade | PESTANAL®, analytical standard |
Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706023
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?