Synonym:
2-
CAS Number 55268-74-1
Empirical Formula (Hill Notation) C19H24N2O2
Molecular Weight 312.41
Beilstein Registry Number 761557
EC Number 259-559-6
MDL number MFCD00058531
PubChem Substance ID 24870350
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Antibiotics, Antibiotics A to Z, |
| grade | VETRANAL™, analytical standard |
| storage temp. | 2-8°C |
VETRANAL is a trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?