Synonym:
1-
CAS Number 70356-09-1
Empirical Formula (Hill Notation) C20H22O3
Molecular Weight 310.39
EC Number 274-581-6
MDL number MFCD00210252
| Related Categories | Analytical Standards, Analytical/Chromatography, Chromatography, Pharmaceutical Secondary Standards, Pharmaceutical Standards More... |
| pharmacopeia traceability | traceable to USP 1045337 |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?