50279 Fluka

Listeria UVM II Supplement

for microbiology

DOWNLOAD MSDS (PDF)

Synonym: Listeria secondary selective enrichment supplement (UVM II)

Properties

Related Categories Media, Microbiology, Supplements More...
grade   for microbiology
shelf life   (limited shelf life, expiry date on the label)
composition   acriflavin hydrochloride, 12.5 mg per vial
  nalidixic, 10.0 mg
storage temp.   2-8°C

Description

Components

Composition:
(per vial, sufficient for 500 mL medium)
Acriflavin hydrchloride 12.5 mg
Nalidixic 10.0 mg

Application

An antimicrobial supplement recommended for selective cultivation of Listeria species.

Price and Availability

Customers Also Viewed

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?