51580 Fluka

Heptadecane

purum, ≥98.0% (GC)

DOWNLOAD MSDS (PDF)
  • CAS Number 629-78-7

  • Linear Formula CH3(CH2)15CH3

  • Molecular Weight 240.47

  •  Beilstein Registry Number 1738898

  •  EC Number 211-108-4

  •  MDL number MFCD00009002

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Acyclic, Alkanes, Aloe Vera, Building Blocks, Chemical Synthesis,
vapor density   8.3 (vs air)
vapor pressure   1 mmHg ( 115 °C)
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.436(lit.)
bp   302 °C(lit.)
mp   20-22 °C(lit.)
density   0.777 g/mL at 25 °C(lit.)

Price and Availability

Customers Also Viewed

Heptadecane

analytical standard

Heptadecane

99%

Heptadecane

analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?