52367 Fluka

Hexadecyltrimethylammonium bromide

for ion pair chromatography, ≥99.0%

DOWNLOAD MSDS (PDF)

Synonym: CTAB, Cetrimonium bromide, Cetyltrimethylammonium bromide, Palmityltrimethylammonium bromide

  • CAS Number 57-09-0

  • Linear Formula CH3(CH2)15N(Br)(CH3)3

  • Molecular Weight 364.45

  •  Beilstein Registry Number 3598189

  •  EC Number 200-311-3

  •  MDL number MFCD00011772

  •  PubChem Substance ID 24874357

  • Popular Documents:  RAMAN FTIR (PDF)  

Properties

Related Categories Acidic Ion Pair Reagents (Anionic), Analytical Reagents, Analytical/Chromatography, Buffers & Reagents, Cationic Detergents,
grade   for ion pair chromatography
description   cationic
assay   ≥99.0% (AT)
  ≥99.0%
mol wt   average mol wt 62,000
aggregation number   170
CMC   0.92
mp   248-251 °C(lit.)
λ   10 % in methanol
UV absorption   λ: 240 nm Amax: 0.04
  λ: 250 nm Amax: 0.03
  λ: 260 nm Amax: 0.02
  λ: 500 nm Amax: 0.02
HLB   11
suitability   corresponds to standard for filter test

Description

Other Notes

Additive for ion-pair chromatography1,2; Ion-pair reagent for TLC3,4

Price and Availability

Customers Also Viewed

Hexadecyltrimethylammonium bromide

BioUltra, for molecular biology, ≥99.0% (AT)

Hexadecyltrimethylammonium bromide

BioUltra, ≥99.0% (AT)

Hexadecyltrimethylammonium bromide

for molecular biology, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?