52370 Fluka

Hexadecyltrimethylammonium bromide

≥96.0% (AT)

DOWNLOAD MSDS (PDF)

Synonym: CTAB, Cetrimonium bromide, Cetyltrimethylammonium bromide, Palmityltrimethylammonium bromide

  • CAS Number 57-09-0

  • Linear Formula CH3(CH2)15N(Br)(CH3)3

  • Molecular Weight 364.45

  •  Beilstein Registry Number 3598189

  •  EC Number 200-311-3

  •  MDL number MFCD00011772

  • Popular Documents:  RAMAN FTIR (PDF)  

Description

Other Notes

Titrant for the potentiometric titration of perchlorate with ionselective electrodes1

Price and Availability

Customers Also Viewed

Hexadecyltrimethylammonium bromide

BioUltra, for molecular biology, ≥99.0% (AT)

Hexadecyltrimethylammonium bromide

for molecular biology, ≥99%

Hexadecyltrimethylammonium bromide

for ion pair chromatography, ≥99.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?