52840 Fluka

1-Hexanol

purum, ≥98.0% (GC)

DOWNLOAD MSDS (PDF)

Synonym: Hexyl alcohol

  • CAS Number 111-27-3

  • Linear Formula CH3(CH2)5OH

  • Molecular Weight 102.17

  •  Beilstein Registry Number 969167

  •  EC Number 203-852-3

  •  MDL number MFCD00002982

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Alcohols, Biochemicals and Reagents, Building Blocks, C2 to C6, Chemical Synthesis,
vapor density   4.5 (vs air)
vapor pressure   1 mmHg ( 25.6 °C)
grade   purum
assay   ≥98.0% (GC)
autoignition temp.   559 °F
refractive index   n20/D 1.418(lit.)
  n20/D 1.418
bp   156-157 °C(lit.)
mp   −52 °C(lit.)
density   0.814 g/mL at 25 °C(lit.)

Price and Availability

Customers Also Viewed

1-Hexanol

analytical standard

1-Hexanol
4714021-Hexanol

anhydrous, ≥99%

1-Hexanol
H133031-Hexanol

reagent grade, 98%

1-Hexanol

ReagentPlus®, ≥99.5% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?