52930 Fluka

1-Hexene

analytical standard

DOWNLOAD MSDS (PDF)
  • CAS Number 592-41-6

  • Linear Formula CH3(CH2)3CH=CH2

  • Molecular Weight 84.16

  •  Beilstein Registry Number 1209240

  •  EC Number 209-753-1

  •  MDL number MFCD00009505

  •  PubChem Substance ID 24877670

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Alpha Sort, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chromatography,
grade   analytical standard
vapor density   3 (vs air)
vapor pressure   155 mmHg ( 21.1 °C)
assay   ≥99.8% (GC)
autoignition temp.   487 °F
shelf life   (limited shelf life, expiry date on the label)
refractive index   n20/D 1.388(lit.)
  n20/D 1.388
bp   60-66 °C(lit.)
density   0.678 g/mL at 25 °C(lit.)

Price and Availability

Proficiency Testing Samples from RTC


PT Samples from RTCRTC has been producing environmental LPTP for more than 20 years and send out over 20,000 PT samples a year, throughout
the world.





Customers Also Viewed

1-Hexene

purum, ≥96.0% (GC)

1-Hexene
2407611-Hexene

≥99%

1-Hexene
3203231-Hexene

97%

1-Hexene
2305451-Hexene

97%

1-Octene

analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?