Synonym:
α-Methyl-d3-
CAS Number 121662-14-4
Empirical Formula (Hill Notation) C13D3H15O2
Molecular Weight 209.30
MDL number MFCD06658737
| Related Categories | APIs & Metabolites, APIs (Active Pharmaceutical Ingredients) & Metabolites, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, |
| grade | analytical standard, for drug analysis |
| isotopic purity | 95 atom % D |
| assay | ≥98.0% (HPLC) |
| shelf life | (limited shelf life, expiry date on the label) |
| mass shift | M+3 |
Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?