Synonym: Δ1-Tetrahydrocannabinol, Δ9-THC
CAS Number 1972-08-3
Empirical Formula (Hill Notation) C21H30O2
Molecular Weight 314.46
EC Number 200-578-6
MDL number MFCD00083207
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Cannabiol, Cannabis, |
| grade | analytical standard, for drug analysis |
| assay | ≥97% (HPLC) |
| shelf life | (limited shelf life, expiry date on the label) |
| drug control | USDEA Schedule I; regulated under CDSA - not available from Sigma-Aldrich Canada |
| concentration | ~1 mg/mL in ethanol |
| suitability | corresponds for H-NMR |
| storage temp. | 2-8°C |
ampoule contains 1.2mL solution
Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?