CAS Number 152120-55-3
Empirical Formula (Hill Notation) C20H18N4O4
Molecular Weight 378.38
Beilstein Registry Number 7779041
MDL number MFCD02092983
PubChem Substance ID 24880275
| Related Categories |
Chemical Biology,
Chemical Synthesis,
Guanidine Protection/Guanidinylatio |
| grade | purum |
| assay | ≥98.0% (TLC) |
| mp | 90-96 °C |
Reagent for solution and solid-phase guanylation of amines to bis-Z-guanidine1†2,3
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?